Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD122
- Clonality:Monoclonal
- Clone:9A2
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Size:100 µg
- Form:liquid
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:YT-1 acute human leukemia cells.
- Cat. No.:102512-996
- Supplier no.:ANC343020
Specifications
About this item
Human CD122 (IL-2 Rbeta) associates with CD25 (IL-2Ralpha) to form a high affinity receptor for IL-2 and is expressed on activated T cells, B cells and monocytes. The cytoplasmic domain of CD122 is involved with signal transduction.
Type: Primary
Antigen: CD122
Clonality: Monoclonal
Clone: 9A2
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG2akappa
Reactivity: Human