Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD105
- Clonality:Monoclonal
- Clone:SN6
- Flow Cytometry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:120 tests
- Form:liquid
- Antigen Synonyms:Endoglin
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Membrane preparation of human B-lineage leukemia cells.
- Cat. No.:102512-972
- Supplier no.:ANC326040
Specifications
About this item
Human CD105 (endoglin) is a homodimeric membrane glycoprotein virtually restricted to endothelial cells and present on some leukemia cells and minor subsets of bone marrow cells. CD105 binds to TGF-beta1 and beta3 with high affinity.
Type: Primary
Antigen: CD105
Clonality: Monoclonal
Clone: SN6
Conjugation: FITC
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human