Mouse IgG2a Isotype Control [clone: RPC 5]
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Immunoglobulin expressed by the mouse plasmacytoma hybridoma RPC 5 is an IgG2a isotype of unknown specificity. It is used as a negative control when working with human cells.
- preservative free
Type: Control
Antigen:
Clonality: Monoclonal
Clone: RPC 5
Conjugation: Preservative Free
Epitope:
Host:
Isotype: Mouse IgG2akappa
Reactivity:
Specifications
- Size:100 µg
- Cat. No.:102512-798
- Clonality:Monoclonal
- Flow Cytometry:Yes
- Shipping Temperature:-20° C
- Antibody Type:Control
- Form:liquid
- Isotype:IgG2a kappa
- Clone:RPC 5
- Storage Temperature:+4°C
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



