To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Control
- Clonality:Monoclonal
- Clone:MOPC31C
- ELISA:Yes
- Flow Cytometry:Yes
- Isotype:IgG1 kappa
- Size:100 µg
- Form:liquid
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Cat. No.:102512-780
- Supplier no.:ANC278040
Specifications
About this item
Immunoglobulin expressed by the mouse plasmacytoma MOPC 31C is an IgG1kappa isotype of unknown specificity. It is used as a negative control when working with human cells.
Type: Control
Antigen:
Clonality: Monoclonal
Clone: MOPC31C
Conjugation: FITC
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: