Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD62E
- Clonality:Monoclonal
- Clone:HAE-1f
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:100 µg
- Form:liquid
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:TNF activated HUVEC.
- Cat. No.:102512-618
- Supplier no.:ANC240020
Specifications
About this item
Human CD62E (E-selectin) is an adhesion molecule expressed on activated endothelium and facilitates recruitment of leukocytes to sights of acute inflammation and immunological events. CD62E has affinity to Lewisx.
Type: Primary
Antigen: CD62E
Clonality: Monoclonal
Clone: HAE-1f
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human