Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD74
- Clonality:Monoclonal
- Clone:M-B741
- Flow Cytometry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Size:120 tests
- Form:liquid
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:RPMI 1788 cell line.
- Cat. No.:102512-580
- Supplier no.:ANC226040
Specifications
About this item
Human CD74 is expressed intracellularly and on the cell surface of MHC Class II positive cells such as B cells, monocytes, macrophages, Langerhans cells and dendritic cells. CD74 has several important functions relating to antigen presentation by MHC Class II molecules.
Type: Primary
Antigen: CD74
Clonality: Monoclonal
Clone: M-B741
Conjugation: FITC
Epitope:
Host:
Isotype: Mouse IgG2a
Reactivity: Human