Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD71
- Clonality:Monoclonal
- Clone:DF1513
- Conjugation:Biotin
- Flow Cytometry:Yes
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:100 µg
- Form:liquid
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Human PBL.
- Cat. No.:102512-568
- Supplier no.:ANC223030
Specifications
About this item
Human CD71 is a type II transmembrane glycoprotein found on activated leukocytes. CD71 binds transferrin and mediates uptake of iron through endocytosis and recycling.
Type: Primary
Antigen: CD71
Clonality: Monoclonal
Clone: DF1513
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human