Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Symbol:CD70
- Clonality:Monoclonal
- Clone:BU69
- Conjugation:Biotin
- ELISA:Yes
- Flow Cytometry:Yes
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Primates,Human
- Size:100 µg
- Form:liquid
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Human WM-1 (Waldenström’s macroglobulinemia) cell line.
- Cat. No.:102512-558
- Supplier no.:ANC222030
Human CD70 is not present on resting lymphocytes. CD70 is expressed after activation on about 20% of T cells and on about 70% of B cells. CD70 is a ligand for CD27 and it is also a member of the TNF gene family.
Type: Primary
Antigen: CD70
Clonality: Monoclonal
Clone: BU69
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human, Primate