Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Symbol:CD69
- Clonality:Monoclonal
- Clone:HP-4B3
- Flow Cytometry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Size:100 µg
- Form:liquid
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:IL-2 activated human NK cells.
- Cat. No.:102512-546
- Supplier no.:ANC221820
Human CD69 is expressed by platelets and mature thymocytes and appears rapidly after activation on T cells, B cells and NK cells. This pattern of cellular CD69 expression has suggested a lymphocyte activation role. Functional studies with antibodies to CD69 also support an activation role for CD69.
- preservative free
Type: Primary
Antigen: CD69
Clonality: Monoclonal
Clone: HP-4B3
Conjugation: Preservative Free
Epitope:
Host:
Isotype: Mouse IgG2a
Reactivity: Human