Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD63
- Clonality:Monoclonal
- Clone:AHN16.1
- Flow Cytometry:Yes
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:100 µg
- Form:liquid
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Human eosinophils.
- Cat. No.:102512-514
- Supplier no.:ANC215820
Specifications
About this item
Human CD63 is expressed on activated platelets and also on monocytes and macrophages. CD63 contains four hydrophobic transmembrane regions and is a member of the tetraspan family. CD63 appears to be involved with cell adhesion and associates with VLA-3 and VLA-6 integrins. In a colon cancer model, it has been correlated to metastatic potential.
- preservative free
Type: Primary
Antigen: CD63
Clonality: Monoclonal
Clone: AHN16.1
Conjugation: Preservative Free
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human