Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Symbol:CD58
- Clonality:Monoclonal
- Clone:TS2
- Flow Cytometry:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:100 µg
- Form:liquid
- Antigen Synonyms:LFA-3
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Human cytotoxic T cells.
- Cat. No.:102512-486
- Supplier no.:ANC210820
Human CD58 is a cell surface glycoprotein with wide distribution on hemopoietic cells and functions as an adhesion molecule mediating a costimulatory signal through CD2. CD58 is attached to the cell surface by either of two forms - transmembrane or GPI.
- preservative free
Type: Primary
Antigen: CD58
Clonality: Monoclonal
Clone: TS2
Conjugation: Preservative Free
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human