Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD45R
- Clonality:Monoclonal
- Clone:351C5
- Conjugation:Biotin
- Flow Cytometry:Yes
- Isotype:IgM
- Reactivity:Human
- Size:100 µg
- Form:liquid
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Peripheral blood mononuclear cells.
- Cat. No.:102512-366
- Supplier no.:ANC195030
Specifications
About this item
Human CD45 is expressed on all cells of hematopoietic origin, except erythrocytes. CD45 is a transmembrane tyrosine phosphatase which can exist in at least nine different isoforms resulting from tissue-specific alternative RNA splicing of exons 4-7 of a single gene coding for the various N-terminal peptide segments. CD45 plays an essential role in lymphocyte activation.
Type: Primary
Antigen: CD45R
Clonality: Monoclonal
Clone: 351C5
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgM
Reactivity: Human