Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD38
- Clonality:Monoclonal
- Clone:AT1
- Conjugation:Biotin
- Flow Cytometry:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:100 µg
- Form:liquid
- Antigen Synonyms:T10|ADP-ribosyl Cyclase 1|Cyclic ADP-ribose Hydrolase 1
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Human T cell tumor cells: CEM.
- Cat. No.:102512-294
- Supplier no.:ANC187030
Specifications
About this item
Human CD38 is a type II transmembrane glycoprotein with bifunctional ectoenzyme activity catalyzing both synthesis and hydrolysis of cyclic ADP ribose. CD38 is involved in lymphocyte activation and adhesion to endothelium and has been identified as a counter-receptor of CD31.
Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: AT1
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human