Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Symbol:CD34
- Clonality:Monoclonal
- Clone:43A1
- Flow Cytometry:Yes
- Isotype:IgG3 kappa
- Reactivity:Primates,Human
- Western Blot:Yes
- Size:100 µg
- Form:liquid
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Human KG-1a tumor cells.
- Cat. No.:102512-252
- Supplier no.:ANC183820
Human CD34 is an unusual glycosylated membrane protein exclusively expressed on immature hematopoietic cells and on endothelial cells. CD34 may have a role in cell adhesion. CD34 has extensive utility for bone marrow transplantation.
- preservative free
Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 43A1
Conjugation: Preservative Free
Epitope:
Host:
Isotype: Mouse IgG3kappa
Reactivity: Human, Primate