Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD31
- Clonality:Monoclonal
- Clone:158-2B3
- Conjugation:Biotin
- Flow Cytometry:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:100 µg
- Form:liquid
- Antigen Synonyms:PECAM-1|GPIIa'|EndoCAM|Plateled Endothelial Cell Adhesion Molecule
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Stimulated human leukocytes.
- Cat. No.:102512-222
- Supplier no.:ANC180030
Specifications
About this item
Human CD31 is an adhesion molecule expressed on platelets, endothelial cells, leukocytes and their bone marrow precursors. CD31 plays a role in homophilic adhesion and heterophilic transendothelial migration.
Type: Primary
Antigen: CD31
Clonality: Monoclonal
Clone: 158-2B3
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human