Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD28
- Clonality:Monoclonal
- Clone:ANC28.1/5D10
- Conjugation:Biotin
- ELISA:Yes
- Flow Cytometry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:liquid
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Recombinant Human CD28 and HPB-MLT human tumor cells.
- Cat. No.:102512-190
- Supplier no.:ANC177030
Specifications
About this item
Human CD28 is an important costimulatory molecule found on all CD4+ T cells and on about half of the CD8+ T cells. T cell activities attributed to CD28 include prevention of anergy, induction of cytokine gene transcription, stabilization of cytokine mRNA and activation of CD8+ cytotoxic T lymphocytes. The ligands for CD28 identified as CD80 (B7-1) and CD86 (B7-2) are immunglobulin superfamily monomeric transmembrane glycoproteins of 60kDa and 80kDa respectively. Anti-CD28 antibodies have been shown to have a strong adjuvant effect when conjugated to an immunogen. Monovalency of the CD28 receptor for its natural ligands is essential to provide costimulation without inducing responses in the absense of TCR engagement.
Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: ANC28.1/5D10
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human