Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Symbol:CD27
- Clonality:Monoclonal
- Clone:M-T271
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:100 µg
- Form:liquid
- Antigen Synonyms:CD27L Receptor|TNFRSF7|T14, Tumor Necrosis Factor Receptor Superfamily Member 7
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:T cell line (from patient with CLL).
- Cat. No.:102512-176
- Supplier no.:ANC176020
Human CD27 is a lymphocyte specific member of the TNF receptor family and is found primarily on peripheral blood T cells and on a subpopulation of B cells and NK cells. The ligand for CD27 is CD70, which is a member of the TNF ligand superfamily.
Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: M-T271
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human