Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD25
- Clonality:Monoclonal
- Clone:7G7B6
- Flow Cytometry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Size:120 tests
- Form:liquid
- Antigen Synonyms:IL-2 Receptor Subunit alpha|p55|IL-2-RA|TAC Antigen
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Stimulated PBMC.
- Cat. No.:102512-160
- Supplier no.:ANC174040
Specifications
About this item
Expression of human CD25 (the alpha subunit of the IL-2 receptor complex) is dramatically upregulated on activated T cells, B cells, NK cells and macrophages. Antibodies to CD25 are being studied for potential use as an immunodirected therapy to selectively attack (kill) CD25 expressing cells.
Type: Primary
Antigen: CD25
Clonality: Monoclonal
Clone: 7G7B6
Conjugation: FITC
Epitope:
Host:
Isotype: Mouse IgG2a
Reactivity: Human