Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Symbol:CD23
- Clonality:Monoclonal
- Clone:BU38
- Flow Cytometry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:120 tests
- Form:liquid
- Antigen Synonyms:BLAST-2|C-type Lectin Domain Family 4 Member J|FcepsilonRII
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Human peripheral myeloma cells.
- Cat. No.:102512-142
- Supplier no.:ANC172040
Human CD23 is a low affinity receptor for IgE (Fc epsilon RII) found primarily on mature B cells and also on eosinophils, monocytes, dendritic cells, platelets and a subset of thymic epithelial cells. CD23 interacts with CD21.
Type: Primary
Antigen: CD23
Clonality: Monoclonal
Clone: BU38
Conjugation: FITC
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human