Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD21
- Clonality:Monoclonal
- Clone:BU33
- Flow Cytometry:Yes
- Isotype:IgG1
- Reactivity:Primates,Human
- Western Blot:Yes
- Size:120 tests
- Form:liquid
- Antigen Synonyms:Complement Receptor Type-2|CR2|Epstein-Barr Virus Receptor
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Human HFB-1 cell line.
- Cat. No.:102512-122
- Supplier no.:ANC170040
Specifications
About this item
Human CD21 is a receptor for complement fragments C3d, C3dg or ic3D and also EBV. CD21 is also a ligand for CD23 and plays a role in IgE synthesis.
Type: Primary
Antigen: CD21
Clonality: Monoclonal
Clone: BU33
Conjugation: FITC
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human, Primate