Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD19
- Clonality:Monoclonal
- Clone:BU12
- Flow Cytometry:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:120 tests
- Form:liquid
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Human EB-4 Burkitt lymphoma cell line.
- Cat. No.:102512-104
- Supplier no.:ANC168050
Specifications
About this item
Human CD19 is a B cell restricted membrane protein expressed from early stages of B cell development and associates with the antigen receptor complex. CD19 is involved in the regulation of B cell proliferation.
Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: BU12
Conjugation: R-PE
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human