Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD18
- Clonality:Monoclonal
- Clone:IB4
- Conjugation:Biotin
- Flow Cytometry:Yes
- Isotype:IgG2a
- Reactivity:Primates,Rabbit,Human
- Size:100 µg
- Form:liquid
- Antigen Synonyms:Integrin beta-2
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Human peripheral blood monocytes.
- Cat. No.:102512-088
- Supplier no.:ANC167030
Specifications
About this item
Human CD18 (beta integrin) forms a heterodimer with either alphaL, alphaM, or alphaX integrins (CD11a, b or c). CD11/CD18 heterodimeric molecules are involved with cell/cell and cell/extracellular adhesion in immune and inflammatory responses.
Type: Primary
Antigen: CD18
Clonality: Monoclonal
Clone: IB4
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG2a
Reactivity: Human, Primate, Rabbit