Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD15
- Clonality:Monoclonal
- Clone:AHN1.1
- Flow Cytometry:Yes
- Isotype:IgM
- Reactivity:Human
- Size:100 µg
- Form:liquid
- Antigen Synonyms:Lewis X
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Human neutrophils.
- Cat. No.:102512-056
- Supplier no.:ANC164820
Specifications
About this item
Human CD15 is expressed by neutrophils, eosinophils and monocytes. Antibodies in this cluster recognize a common terminal pentasaccharide found on cell surface glycoproteins and glycolipids.
- preservative free
Type: Primary
Antigen: CD15
Clonality: Monoclonal
Clone: AHN1.1
Conjugation: Preservative Free
Epitope:
Host:
Isotype: Mouse IgM
Reactivity: Human