Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Symbol:CD14
- Clonality:Monoclonal
- Clone:UCHM1
- Conjugation:Biotin
- Flow Cytometry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Size:100 µg
- Form:liquid
- Antigen Synonyms:Myeloid Cell-specific Leucine-rich Glycoprotein
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Human thymocytes/Sezary T cells.
- Cat. No.:102512-048
- Supplier no.:ANC163030
Human CD14 is strongly expressed on myelomonocyte lineage cells including monocytes, macrophages and a subset of granulocytes. CD14 is a high affinity receptor for complexes of lipopolysaccharide and lipopolysaccharide binding protein. A soluble form of CD14 also binds to LPS and LBP.
Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: UCHM1
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG2a
Reactivity: Human