Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Symbol:CD11B
- Clonality:Monoclonal
- Clone:ICRF44
- Flow Cytometry:Yes
- Isotype:IgG1
- Reactivity:Primates,Human
- Size:100 µg
- Form:liquid
- Antigen Synonyms:Cell Surface Glycoprotein MAC-1 Subunit alpha|Integrin alpha-M|Leukocyte Adhesion Receptor MO1
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Human monocytes.
- Cat. No.:102512-016
- Supplier no.:ANC159820
Human CD11b (alpha M integrin) complexes with CD18 (beta 2 integrin) to form the complement receptor type 3 (CR3) heterodimer which binds to ICAM-1, fibrinogen, C3b and coagulation factor X. CD11b expression is restricted to leukocytes, mainly to myeloid and NK cells.
- preservative free
Type: Primary
Antigen: CD11B
Clonality: Monoclonal
Clone: ICRF44
Conjugation: Preservative Free
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human, Primate