Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD11A
- Clonality:Monoclonal
- Clone:38
- Flow Cytometry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western Blot:Yes
- Size:120 tests
- Form:liquid
- Antigen Synonyms:Leukocyte Adhesion Glycoprotein LFA-1 alpha Chain|Integrin alpha-L|Lymphocyte Antigen 15|Ly-15
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Human monocytes.
- Cat. No.:102512-012
- Supplier no.:ANC158050
Specifications
About this item
Human CD11a (alpha L integrin) complexes with CD18 (beta 2 integrin) to form the LFA-1 heterodimer adhesion molecule which binds to three ICAM’s 1, 2, and 3, also designated CD54, CD102 and CD50 respectively. LFA-1 is expressed on lymphocytes, monocytes and neutrophils and plays a role in cell-cell adhesion.
Type: Primary
Antigen: CD11A
Clonality: Monoclonal
Clone: 38
Conjugation: R-PE
Epitope:
Host:
Isotype: Mouse IgG2a
Reactivity: Human