To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD10
- Clonality:Monoclonal
- Clone:SN5c
- Flow Cytometry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:120 tests
- Form:liquid
- Antigen Synonyms:Enkephalinase|CALLA|Skin Fibroblast Elastase|Atriopeptidase|SFE|Neprilysin
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Membrane preparation of human B-lineage leukemia cells.
- Cat. No.:102512-000
- Supplier no.:ANC157040
Specifications
About this item
Human CD10 is a well known marker for common acute lymphoblastic leukemia antigen (CALLA). CD10 is expressed on normal early B cells in bone marrow and is also on fetal thymocytes. CD10 expression is lost as the T and B lineage cells mature and only appears again on activated germinal center B cells. CD10 is present on neutrophils. CD10 is a neutral endopeptidase and hydrolyzes peptide bonds on the amino side of hydrophobic amino acids.
Type: Primary
Antigen: CD10
Clonality: Monoclonal
Clone: SN5c
Conjugation: FITC
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human