To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD9
- Clonality:Monoclonal
- Clone:SN4
- Flow Cytometry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:120 tests
- Form:liquid
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Membrane antigen preparation of NALM-6.
- Cat. No.:102511-992
- Supplier no.:ANC156050
Specifications
About this item
Human CD9 is strongly expressed on platelets, on early B cells and on activated T cells. CD9 is a member of the tetraspan family crossing the membrane four times. Both the N-terminal and C-terminal regions are on the cytoplasmic side of the plasma membrane. Studies indicate CD9 may be involved with calcium mobilization.
Type: Primary
Antigen: CD9
Clonality: Monoclonal
Clone: SN4
Conjugation: R-PE
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human