Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD49b
- Clonality:Monoclonal
- Clone:HAS6
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Size:100 µg
- Form:liquid
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Human cultured keratinocytes.
- Cat. No.:102511-974
- Supplier no.:ANC155020
Specifications
About this item
Human CD49b is an integrin alpha2 subunit that forms a heterodimer with beta1 integrin and functions as an adhesion molecule.
Type: Primary
Antigen: CD49b
Clonality: Monoclonal
Clone: HAS6
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG2akappa
Reactivity: Human