Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Symbol:CD8
- Clonality:Monoclonal
- Clone:UCHT4
- ELISA:Yes
- Flow Cytometry:Yes
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western Blot:Yes
- Size:120 tests
- Form:liquid
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Human thymocytes/Sezary T cells.
- Cat. No.:102511-958
- Supplier no.:ANC153050
Human CD8 is a heterodimeric protein consisting of an alpha and beta chain, each about 33kDa. CD8 is expressed on most thymocytes and on about one third of peripheral blood T cells. CD8 is a co-receptor involved in antigen recognition and binds to the alpha2 and alpha3 domain of MHC Class I molecules.
Type: Primary
Antigen: CD8
Clonality: Monoclonal
Clone: UCHT4
Conjugation: R-PE
Epitope:
Host:
Isotype: Mouse IgG2a
Reactivity: Human