Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD7
- Clonality:Monoclonal
- Clone:3A1E
- Conjugation:Biotin
- Flow Cytometry:Yes
- Isotype:IgG2b
- Reactivity:Human
- Size:100 µg
- Form:liquid
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Human HSB2-T cells.
- Cat. No.:102511-944
- Supplier no.:ANC152030
Specifications
About this item
Human CD7 is one of the earliest T lineage markers and is found on most thymocytes and peripheral blood T cells. The function of CD7 is not yet fully known, although cross-linking of CD7 with antibodies induces a T cell transmembrane calcium flux and CD7 expression is induced by ionomycin.
Type: Primary
Antigen: CD7
Clonality: Monoclonal
Clone: 3A1E
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG2b
Reactivity: Human