Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Symbol:CD6
- Clonality:Monoclonal
- Clone:3F7B6
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:liquid
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Human rheumatoid synovial T cell line (ST-1).
- Cat. No.:102511-930
- Supplier no.:ANC151020
Human CD6 is expressed on peripheral blood T cells, most thymocytes and a subset of B cells. CD6 is similar to CD5 and contains three cysteine-rich domains with structural homology to the macrophage scavenger receptor. CD6 is involved in T cell activation and its ligand was recently identified.
Type: Primary
Antigen: CD6
Clonality: Monoclonal
Clone: 3F7B6
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human