Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD4
- Clonality:Monoclonal
- Clone:M-T441
- ELISA:Yes
- Flow Cytometry:Yes
- Isotype:IgG2b
- Reactivity:Baboon,Human
- Size:100 µg
- Form:liquid
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:P815 cells transfected with human CD4.
- Cat. No.:102511-910
- Supplier no.:ANC148820
Specifications
About this item
Human CD4 is a cell surface glycoprotein of 55kDa expressed on most thymocytes, on about two thirds of peripheral T cells and on some monocyte macrophage lineage cells. CD4 interacts with MHC Class II molecules in an accessory role during foreign antigen recognition by T cells. The extracellular portion of CD4 is an array of four Ig-like domains. The N-terminal region of CD4 is a receptor for the HIV-1 viral protein gp120.
- preservative free
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: M-T441
Conjugation: Preservative Free
Epitope:
Host:
Isotype: Mouse IgG2b
Reactivity: Baboon, Human