Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Symbol:CD1c
- Clonality:Monoclonal
- Clone:M241
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Isotype:IgG1 kappa
- Reactivity:Rabbit,Human
- Size:100 µg
- Form:liquid
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Human T cell line Molt 4.
- Cat. No.:102511-882
- Supplier no.:ANC146020
The human CD1 genetic locus consists of five closely related genes. Three of the gene products (CD1a, CD1b and CD1c) have been officially clustered using antibodies and are expressed on cortical thymocytes, a subset of B cells, and on most professional antigen presenting cells. CD1 molecules have a specialized lipid antigen-presenting function and it is unlikely that they present conventional peptide antigens. CD1 may also be involved in costimulation.
Type: Primary
Antigen: CD1c
Clonality: Monoclonal
Clone: M241
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human, Rabbit