Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD1a
- Clonality:Monoclonal
- Clone:CB-T6
- Flow Cytometry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:liquid
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Human Jurkat tumor cells.
- Cat. No.:102511-874
- Supplier no.:ANC145820
Specifications
About this item
The human CD1 genetic locus consists of five closely related genes. Three of the gene products (CD1a, CD1b and CD1c) have been officially clustered using antibodies and are expressed on cortical thymocytes, a subset of B cells, and on most professional antigen presenting cells. CD1a+/CD11c+ dendritic cells are direct precursors of Langerhans cells in peripheral blood. CD1 molecules have a specialized lipid antigen-presenting function.
- preservative free
Type: Primary
Antigen: CD1a
Clonality: Monoclonal
Clone: CB-T6
Conjugation: Preservative Free
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human