Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD3
- Clonality:Monoclonal
- Clone:UCHT1
- Flow Cytometry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:120 tests
- Form:liquid
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Human thymocytes/Sezary T cells.
- Cat. No.:102511-866
- Supplier no.:ANC144060
Specifications
About this item
The human CD3/T cell receptor (TcR) complex is made up of the CD3 proteins (gamma, delta, 2 epsilon) and teh zeta-chain in association with either alpha/beta or gamma/delta proteins of the TcR. The TcR recognizes antigens in association with MHC molecules after which protein chains of the CD3 complex mediate activation signals triggered by TcR antigen binding. CD3 is expressed on greater than 95% of circulating human peripheral T cells.
Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: UCHT1
Conjugation: APC
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human