Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Symbol:CD2
- Clonality:Monoclonal
- Clone:1E7E8
- Flow Cytometry:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:120 tests
- Form:liquid
- Antigen Synonyms:Rosette Receptor|LFA-3 Receptor|LFA-2|Erythrocyte Receptor
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Human rheumatoid synovial T cell line (ST-1).
- Cat. No.:102511-854
- Supplier no.:ANC143050
Human CD2 is a T-lineage glycoprotein with roles in cell-cell adhesion and the transduction of activation signals. Multiple ligands for CD2 include CD58 (LFA-3), CD59, and CD48. CD2 may play a role in regulating T cell anergy.
Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: 1E7E8
Conjugation: R-PE
Epitope:
Host:
Isotype: Mouse IgG2akappa
Reactivity: Human