Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Secondary
- Antigen Symbol:IgM
- Clonality:Monoclonal
- Clone:UCHB1
- ELISA:Yes
- Flow Cytometry:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:100 µg
- Form:liquid
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Prolymphocytic leukemic B cells.
- Cat. No.:102511-828
- Supplier no.:ANC141820
Specifications
About this item
Human surface membrane bound IgM complexes with a heterodimer of transmembrane proteins of Igalpha and Igbeta (CD79a and CD79b) to form the B cell antigen receptor (BCR). The BCR complex plays an important role in B cell development and activation. The transmembrane region of mIgM is essential for functional BCR complex assembly.
- preservative free
Type: Secondary
Antigen: IgM
Clonality: Monoclonal
Clone: UCHB1
Conjugation: Preservative Free
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human