Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Symbol:MHC1
- Clonality:Monoclonal
- Clone:3F10
- ELISA:Yes
- Flow Cytometry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Size:120 tests
- Form:liquid
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Human HSB-2 T cell line.
- Cat. No.:102511-762
- Supplier no.:ANC121040
Human MHC Class I (HLA-A, B & C) molecules are expressed on nearly all nucleated cells. MHC Class I molecules consist of heterodimers of a highly polymorphic transmembrane alpha chain and an invariant beta chain. MHC Class I molecules bind to endogenously synthesized peptides of about 8-10 amino acids which are presented to CD8+ T cells. Evidence indicates exogenously derived peptides are also presented by MHC Class I molecules.
Type: Primary
Antigen: MHC1
Clonality: Monoclonal
Clone: 3F10
Conjugation: FITC
Epitope:
Host:
Isotype: Mouse IgG2a
Reactivity: Human