Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Symbol:TCR
- Clonality:Monoclonal
- Clone:Jovi-3
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Isotype:IgG2a lambda
- Reactivity:Human
- Size:100 µg
- Form:liquid
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Human HA1.7 TCR beta chain expressed on transgenic mouse cells.
- Cat. No.:102511-738
- Supplier no.:ANC102020
Human T cell receptor (TCR) is a T cell surface expressed heterodimer of alpha/beta or gamma/delta chains. TCR’s recognize antigen peptides bound to MHC molecules forming the basis of antigen specific response by T cells.
Type: Primary
Antigen: TCR
Clonality: Monoclonal
Clone: Jovi-3
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG2alambda
Reactivity: Human