To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:ANGPT-2
- Clone:Angy-2-1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:CHO cells
- Isotype:IgG2b lambda
- Reactivity:Human,Mouse
- Size:100 µg
- Form:liquid
- Gene ID:O15123
- Antigen Synonyms:Agpt2|Ang-2|Angpt2|Ang2
- Storage Temperature:–20 °C
- Concentration:1mg/ml
- Shipping Temperature:-20° C
- Immunogen:Recombinant human angiopoietin-2.
- Cat. No.:102978-642
- Supplier no.:AG27B0016C100
Angiopoietin-1 (Ang-1) and Angiopoietin-2 (Ang-2) are closely related secreted ligands which bind with similar affinity to Tie-2. Tie-2 and angiopoietins have been shown to play critical roles in embryogenic angiogenesis and in maintaining the integrity of the adult vasculature. Ang-1 activates Tie-2 signaling on endothelial cells to promote chemotaxis, cell survival, cell sprouting, vessel growth and stabilization. Ang-2 has been identified as a secreted protein ligand of Tie-2 and has alternatively been reported to be an antagonist for Ang-1 induced Tie-2 signaling as well as an agonist for Tie-2 signaling, depending on the cell context. anti-Angiopoietin-2, mAb (rec.) (blocking) (Angy-2-1) is an antibody developed by antibody phage display technology using a human naive antibody gene library. These libraries consist of scFv (single chain fragment variable) composed of VH (variable domain of the human immunoglobulin heavy chain) and VL (variable domain of the human immunoglobulin light chain) connected by a polypeptide linker. The antibody fragments are displayed on the surface of filamentous bacteriophage (M13). This scFv was selected by affinity selection on antigen in a process termed panning. Multiple rounds of panning are performed to enrich for antigen-specific scFv-phage. Monoclonal antibodies are subsequently identified by screening after each round of selection. The selected monoclonal scFv is cloned into an appropriate vector containing a Fc portion of interest and then produced in mammalian cells to generate an IgG like scFv-Fc fusion protein.
Type: Primary
Antigen: ANGPT-2
Clonality: Recombinant
Clone: Angy-2-1
Conjugation: Unconjugated
Epitope:
Host: CHO cells
Isotype: Mouse IgG2blambda.
Reactivity: Human, Mouse