Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Symbol:IL-33
- Clone:Bondy-1-1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:HEK293
- Isotype:IgG2b
- Reactivity:Mouse
- Size:100 µg
- Form:liquid
- Gene ID:Q8BVZ5
- Antigen Synonyms:IL-1F11|NF-HEV|Interleukin-33
- Storage Temperature:–20 °C
- Concentration:1mg/ml
- Shipping Temperature:-20° C
- Immunogen:Mouse recombinant IL-33.
- Cat. No.:102980-006
- Supplier no.:AG27B0013C100
Interleukin-33 (IL-33; HF-NEV; IL-1F11), a member of the IL-1 family of cytokines, is expressed by many cell types following pro-inflammatory stimulation and is thought to be released upon cell lysis. IL-33 binds to and signals through ST2 (IL-1R1) and its stimulation recruits MYD88, IRAK, IRAK4 and TRAF6, followed by phosphorylation of ERK1 (MAPK3)/ERK2 (MAPK1), p38 (MAPK14) and JNK. The ability of IL-33 to target numerous immune cell types, like Th2-like cells, mast cells and B1 cells, and to induce cytokine and chemokine production underlines its potential in influencing the outcome of a wide range of diseases, such as arthritis, asthma, atopic allergy & anaphylaxis, cardiovascular disease/atherosclerosis, nervous system diseases and sepsis. Anti-IL-33, mAb (recombinant) (blocking) (Bondy-1-1) is an antibody developed by antibody phage display technology using a human naive antibody gene library. These libraries consist of scFv (single chain fragment variable) composed of VH (variable domain of the human immunoglobulin heavy chain) and VL (variable domain of the human immunoglobulin light chain) connected by a polypeptide linker. The antibody fragments are displayed on the surface of filamentous bacteriophage (M13). This scFv was selected by affinity selection on antigen in a process termed panning. Multiple rounds of panning are performed to enrich for antigen-specific scFv-phage. Monoclonal antibodies are subsequently identified by screening after each round of selection. The selected monoclonal scFv is cloned into an appropriate vector containing a Fc portion of interest and then produced in mammalian cells to generate an IgG like scFv-Fc fusion protein.
Type: Primary
Antigen: IL-33
Clonality: Recombinant
Clone: Bondy-1-1
Conjugation: Unconjugated
Epitope:
Host: Purified from HEK 293 cell culture supernatant.
Isotype: Mouse IgG2b
Reactivity: Mouse