Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:ASC
- Clonality:Polyclonal
- Conjugation:ATTO 647N
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Size:100 µg
- Form:liquid
- Gene ID:Q9ULZ3
- Antigen Synonyms:Target of Methylation-induced Silencing 1|CARD5|Apoptosis-associated Speck-like Protein Containing CARD|Pycard|Caspase Recruitment Domain-containing Protein 5|TMS1
- Storage Temperature:+4°C
- Concentration:0.5mg/ml
- Shipping Temperature:-20° C
- Immunogen:Synthetic peptide corresponding to aa at the N-terminal human Asc.
- Cat. No.:102979-932
- Supplier no.:25B0006TSC100
Specifications
About this item
Asc promotes caspase-mediated apoptosis. This proapoptotic activity is mediated predominantly through the activation of caspase-9. It is a component of the inflammasome, a protein complex which also includes NLRP3/NALP3, CARD8 and CASP1 and whose function is the activation of proinflammatory caspases.
Type: Primary
Antigen: ASC
Clonality: Polyclonal
Clone:
Conjugation: ATTO 647N
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse