Anti-EIF2B5 Rabbit Polyclonal Antibody
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
This gene encodes one of five subunits of eukaryotic translation initiation factor 2B (EIF2B), a GTP exchange factor for eukaryotic initiation factor 2 and an essential regulator for protein synthesis. Mutations in this gene and the genes encoding other EIF2B subunits have been associated with leukoencephalopathy with vanishing white matter.
For WB starting dilution is: 1:1000 For IHC-P starting dilution is: 1:10~50
Type: Primary
Antigen: EIF2B5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human, Mouse
Specifications
- Size:400 ul
- Clonality:Polyclonal
- Modification:None
- Molecular Weight:80 kDa
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Epitope:Amino Acid Range = 36-65
- Form:Liquid
- Storage Buffer:Supplied in PBS with 0.09% (W/V) sodium azide.
- Host:Rabbit
- Concentration:0.5 mg/mL
- Cat. No.:76008-796
- Reactivity:Human|Mouse
- Western Blot:Yes
- Shipping Temperature:4 °C
- Antibody Type:Primary
- Antigen Symbol:EIF2B5
- Gene ID:Q13144
- Immunogen:This EIF2B5 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 36-65 amino acids from the N-terminal region of human EIF2B5.
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen Synonyms:EIF2B5
- Antigen Name:Translation initiation factor eIF-2B subunit epsilon
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



