Specifications
- Antibody Type:Primary
- Antigen Name:Cadherin-19
- Antigen Symbol:CDH19
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:400 ul
- Epitope:Amino Acid Range = 613-640
- Form:Liquid
- Gene ID:Q9H159
- Antigen Synonyms:CDH19
- Modification:None
- Storage Buffer:Supplied in PBS with 0.09% (W/V) sodium azide.
- Molecular Weight:87 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.5 mg/mL
- Shipping Temperature:4 °C
- Immunogen:This CDH19 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 613-640 amino acids from the C-terminal region of human CDH19.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76012-380
Specifications
About this item
This gene is a type II classical cadherin from the cadherin superfamily and one of three cadherin 7-like genes located in a cluster on chromosome 18. The encoded membrane protein is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Type II (atypical) cadherins are defined based on their lack of a HAV cell adhesion recognition sequence specific to type I cadherins. Since disturbance of intracellular adhesion is a prerequisite for invasion and metastasis of tumor cells, cadherins are considered prime candidates for tumor suppressor genes.
For WB starting dilution is: 1:1000 For IHC-P starting dilution is: 1:10~50
Type: Primary
Antigen: CDH19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human