- Antibody Type:Primary
- Antigen Name:Lactadherin
- Antigen Symbol:MFGE8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:400 ul
- Epitope:Amino Acid Range = 316-345
- Form:Liquid
- Gene ID:Q08431
- Antigen Synonyms:MFGE8
- Modification:None
- Storage Buffer:Supplied in PBS with 0.09% (W/V) sodium azide.
- Molecular Weight:43 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.5 mg/mL
- Shipping Temperature:4 °C
- Immunogen:This HMFG antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 316-345 amino acids from the C-terminal region of human HMFG.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76012-306
HMFG plays an important role in the maintenance of intestinal epithelial homeostasis and the promotion of mucosal healing. Promotes VEGF-dependent neovascularization (By similarity). Contributes to phagocytic removal of apoptotic cells in many tissues. Specific ligand for the alpha-v/beta-3 and alpha-v/beta-5 receptors. Also binds to phosphatidylserine-enriched cell surfaces in a receptor-independent manner. Zona pellucida-binding protein which may play a role in gamete interaction. Binds specifically to rotavirus and inhibits its replication. Medin is the main constituent of aortic medial amyloid.
For WB starting dilution is: 1:1000 For FACS starting dilution is: 1:10~50
Type: Primary
Antigen: MFGE8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human