Anti-MGAT4C Rabbit Polyclonal Antibody
Catalog # 76011-002
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Alpha-1,3-mannosyl-glycoprotein 4-beta-N-acetylglucosaminyltransferase C
- Antigen Symbol:MGAT4C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:400 ul
- Epitope:Amino Acid Range = 303-332
- Form:Liquid
- Gene ID:Q9UBM8
- Antigen Synonyms:MGAT4C
- Modification:None
- Storage Buffer:Supplied in PBS with 0.09% (W/V) sodium azide.
- Molecular Weight:56 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.5 mg/mL
- Shipping Temperature:4 °C
- Immunogen:This MGAT4C antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 303-332 amino acids from the C-terminal region of human MGAT4C.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76011-002
Specifications
About this item
Glycosyltransferase that participates in the transfer of N-acetylglucosamine (GlcNAc) to the core mannose residues of N-linked glycans. Catalyzes the formation of the GlcNAcbeta1-4 branch on the GlcNAcbeta1-2Manalpha1-3 arm of the core structure of N-linked glycans. Essential for the production of tri-and tetra-antennary N-linked sugar chains (By similarity). Does not catalyze the transfer of GlcNAc to the Manalpha1-6 arm to form GlcNAcBeta1-4Manalpha1-6 linkage ('GnT-VI' activity).
For WB starting dilution is: 1:1000 For IHC-P starting dilution is: 1:10~50
Type: Primary
Antigen: MGAT4C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human