Specifications
- Antibody Type:Primary
- Antigen Name:Catenin delta-2
- Antigen Symbol:CTNND2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:400 ul
- Epitope:Amino Acid Range = 1131-1159
- Form:Liquid
- Gene ID:Q9UQB3
- Antigen Synonyms:CTNND2
- Modification:None
- Storage Buffer:Supplied in PBS with 0.09% (W/V) sodium azide.
- Molecular Weight:133 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.5 mg/mL
- Shipping Temperature:4 °C
- Immunogen:This CTNND2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 1131-1159 amino acids from the C-terminal region of human CTNND2.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76011-984
Specifications
About this item
This gene encodes an adhesive junction associated protein of the armadillo/beta-catenin superfamily and is implicated in brain and eye development and cancer formation. The protein encoded by this gene promotes the disruption of E-cadherin based adherens junction to favor cell spreading upon stimulation by hepatocyte growth factor. This gene is overexpressed in prostate adenocarcinomas and is associated with decreased expression of tumor suppressor E-cadherin in this tissue. This gene resides in a region of the short arm of chromosome 5 that is deleted in Cri du Chat syndrome.
For WB starting dilution is: 1:1000 For IHC-P starting dilution is: 1:10~50
Type: Primary
Antigen: CTNND2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human, Mouse