Specifications
- Antibody Type:Primary
- Antigen Name:Selenoprotein N
- Antigen Symbol:SEPN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:400 ul
- Epitope:Amino Acid Range = 417-445
- Form:Liquid
- Gene ID:Q9NZV5
- Antigen Synonyms:SEPN1
- Modification:None
- Storage Buffer:Supplied in PBS with 0.09% (W/V) sodium azide.
- Molecular Weight:66 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.5 mg/mL
- Shipping Temperature:4 °C
- Immunogen:This SEPN1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 417-445 amino acids from the C-terminal region of human SEPN1.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76011-914
Specifications
About this item
This gene encodes a selenoprotein, which contains a selenocysteine (Sec) residue at its active site. The selenocysteine is encoded by the UGA codon that normally signals translation termination. The 3' UTR of selenoprotein genes have a common stem-loop structure, the sec insertion sequence (SECIS), that is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. Mutations in this gene cause the classical phenotype of multiminicore disease and congenital muscular dystrophy with spinal rigidity and restrictive respiratory syndrome. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
For IHC-P starting dilution is: 1:10~50 For WB starting dilution is: 1:1000
Type: Primary
Antigen: SEPN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human