Specifications
- Antibody Type:Primary
- Antigen Name:Kelch repeat and BTB domain-containing protein 13
- Antigen Symbol:KBTBD13
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:400 ul
- Epitope:Amino Acid Range = 409-438
- Form:Liquid
- Gene ID:C9JR72
- Antigen Synonyms:KBTBD13
- Modification:None
- Storage Buffer:Supplied in PBS with 0.09% (W/V) sodium azide.
- Molecular Weight:49 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.44 mg/mL
- Shipping Temperature:4 °C
- Immunogen:This hCG_1645727 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 409-438 amino acids from the C-terminal region of human hCG_1645727.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76011-584
Specifications
About this item
The gene belongs to a family of genes encoding proteins containing a BTB domain and several kelch repeats. The BTB domain functions as a protein-protein interaction module, which includes an ability to self-associate or to interact with non-BTB domain-containing proteins. The kelch motif typically occurs in groups of five to seven repeats, and has been found in proteins with diverse functions. Known functions of these family members include transcription regulation, ion channel tetramerization and gating, protein ubiquitination or degradation, and cytoskeleton regulation. The exact function of this family member has yet to be determined.
For IHC-P starting dilution is: 1:10~50 For WB starting dilution is: 1:1000
Type: Primary
Antigen: KBTBD13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human